Cole-Parmer Sodium Hydroxide (Pellets) ACS, 10 K

Representative image only.
Cole-Parmer Item # EW-88404-72
Reagents and solutions to meet your lab and research needs
  • Quality and consistency your lab can count on

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • ClassReagent Acids and Bases
  • ChemistrySolutions;Analytical Reagents
  • FormSolid
  • Weight (g)10000
  • PackagingFiber Drum
  • DOT InformationSodium hydroxide, solid; Class 8, Packing Group II
  • UN NumberUN1823
  • CAS Number1310-73-2
  • Shelf Life (days)60
  • DescriptionSodium Hydroxide (Pellets) ACS, 10 K

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat