Cole-Parmer Sodium Lauryl Sulfate (Sodium Dodecyl Sulfate), Reagent Grade, 25 g

Representative image only.
Fisher Chemical Item # EW-88404-79
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs

SPECS & DESCRIPTION

Specifications & Description

  • ClassReagent Salts
  • ChemistrySolutions
  • FormSolid
  • Weight (g)25
  • PackagingPouch
  • DOT InformationFlammable solids, organic, nos
  • CAS Number151-21-3
  • UN NumberUN1325
  • Shelf Life (days)60
  • DescriptionSodium Lauryl Sulfate (Sodium Dodecyl Sulfate), Reagent Grade, 25 g

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Ideal for protein extraction procedures and SDS polyacrylamide gels.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat