Cole-Parmer Sodium Molybdate Dihydrate ACS; 100 g

Representative image only.
Fisher Chemical Item # EW-88404-80
Reagents and solutions to meet your lab and research needs
  • Assay - 99.5-103.0%
  • Insoluble Matter - ≤0.005%
  • Chloride - ≤0.005%
  • Phosphate - ≤5ppm
  • Sulfate - ≤0.015%
  • Ammonium - ≤0.001%
  • Heavy Metals (as Pb) - ≤5ppm
  • pH of a 5% Solution at 25°C - 7.0 to 10.5
  • Iron - ≤0.001%

SPECS & DESCRIPTION

Specifications & Description

  • ClassReagent Salts
  • ChemistryAnalytical Reagents;Solutions
  • FormSolid
  • Weight (g)100
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number10102-40-6
  • Shelf Life (days)120
  • DescriptionSodium Molybdate Dihydrate ACS, 100 g

Key Features

  • Assay - 99.5-103.0%
  • Insoluble Matter - ≤0.005%
  • Chloride - ≤0.005%
  • Phosphate - ≤5ppm
  • Sulfate - ≤0.015%
  • Ammonium - ≤0.001%
  • Heavy Metals (as Pb) - ≤5ppm
  • pH of a 5% Solution at 25°C - 7.0 to 10.5
  • Iron - ≤0.001%

More About this Item

Quality and consistency your lab can count on.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat