Cole-Parmer Sodium Peroxide, ACS Grade, 125 mL

Representative image only.
Cole-Parmer Item # EW-88404-90

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • ChemistryAnalytical Reagents;Solutions
  • Volume (mL)125
  • PackagingMetal Can
  • DOT InformationSodium Peroxide
  • CAS Number1313-60-6
  • UN NumberUN1504
  • Shelf Life (months)24
  • DescriptionSodium Peroxide, ACS Grade, 125 mL

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat