Cole-Parmer Essentials Solid Black Rubber Stoppers, Standard Size 13.5; 3/Pk
Representative image only.
Cole-Parmer Essentials Item # EW-62990-46
$79.10USD / Pkg of 3
BUY MORE & SAVE
  • 1-5$79.10USD / Pkg of 3
  • 6-29$71.19USD / Pkg of 3
  • 30-119$67.24USD / Pkg of 3
  • 120+$63.28USD / Pkg of 3
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number13.5
  • Top Outside Diameter (OD) (mm)75
  • Bottom Outside Diameter (OD) (mm)62
  • Height (mm)35
  • ColorBlack
  • MaterialRubber
  • DescriptionSolid Black Rubber Stoppers, Standard Size 13.5; 3/Pk
More About this Item
Black rubber stoppers stay pliant over a long period and withstand laboratory conditions without hardening. Stoppers retain resiliency even after repeated autoclaving. Each stopper has its size in raised figures on top. Contain some sulfur. Autoclavable.