Cole-Parmer Essentials Solid Black Rubber Stoppers, Standard Size 15; 1/Pk
Representative image only.
Cole-Parmer Essentials Item # EW-62990-50
$87.70USD / Pkg of 1
BUY MORE & SAVE
  • 1-5$87.70USD / Pkg of 1
  • 6-29$78.93USD / Pkg of 1
  • 30-119$74.55USD / Pkg of 1
  • 120+$70.16USD / Pkg of 1
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number15
  • Top Outside Diameter (OD) (mm)103
  • Bottom Outside Diameter (OD) (mm)83
  • Height (mm)39
  • ColorBlack
  • MaterialRubber
  • DescriptionSolid Black Rubber Stoppers, Standard Size 15; 1/Pk
More About this Item
Black rubber stoppers stay pliant over a long period and withstand laboratory conditions without hardening. Stoppers retain resiliency even after repeated autoclaving. Each stopper has its size in raised figures on top. Contain some sulfur. Autoclavable.