Cole-Parmer Essentials Solid Green Neoprene Stoppers, Standard Size 14; 1/Pk
Representative image only.
Cole-Parmer Essentials Item # EW-62991-48
$81.20USD / Pkg of 1
BUY MORE & SAVE
  • 1-5$81.20USD / Pkg of 1
  • 6-29$73.08USD / Pkg of 1
  • 30-119$69.02USD / Pkg of 1
  • 120+$64.96USD / Pkg of 1
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number14
  • Top Outside Diameter (OD) (mm)90
  • Bottom Outside Diameter (OD) (mm)75
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 14; 1/Pk
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.