Cole-Parmer Essentials Solid Green Neoprene Stoppers, Standard Size 15; 1/Pk
Representative image only.
Cole-Parmer Essentials Item # EW-62991-50
$88.40USD / Pkg of 1
BUY MORE & SAVE
  • 1-5$88.40USD / Pkg of 1
  • 6-29$79.56USD / Pkg of 1
  • 30-119$75.14USD / Pkg of 1
  • 120+$70.72USD / Pkg of 1
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number15
  • Top Outside Diameter (OD) (mm)103
  • Bottom Outside Diameter (OD) (mm)83
  • Height (mm)39
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 15; 1/Pk
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.