Cole-Parmer Solid SS Pan for 0.75 gal and SS 1 gal Cleaners
Representative image only.
Cole-Parmer Item # EW-08851-41
Achieve optimal cleaning results
  • Supports items that require indirect cleaning
  • Prevents damage to transducers
  • 1 year warranty
PRODUCT OPTIONS
$105.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Material304 SS
  • DescriptionSolid SS Pan for 0.75 gal and SS 1 gal Cleaners
  • Warranty1 Year
1 Year Warranty
Key Features
  • Supports items that require indirect cleaning
  • Prevents damage to transducers
  • 1 year warranty
More About this Item
Made of 304 stainless steel, these solid insert trays can be used to support items that require indirect cleaning. Ultrasonic activity is greatest about an inch from the bottom of the tank, so suspending your items provides the best cleaning possible and helps prevent damage to the transducers.
Never use ultrasonic cleaners with flammable solvents. A cleaning solution must be used with these cleaners. Use an insert tray or beaker positioning cover and beaker with these ultrasonic cleaners. Ultrasonic acticity is greatest starting about an inch from the bottom of the tank.