Cole-Parmer Solution Grounded pH Probe, DJ/PPS/100Ohm RTD w/spade lugs/BNC
Representative image only.
Cole-Parmer Item # EW-27018-02
Industrial pH/ORP Electrodes for Chemical Property Testing
  • Get longer life with PTFE double junctions
  • Ideal for harsh conditions
  • 1 year warranty
PRODUCT OPTIONS
$396.00USD / Each
BUY MORE & SAVE
  • 1-5$396.00USD / Each
  • 6-11$356.40USD / Each
  • 12-23$336.60USD / Each
  • 24+$316.80USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialRyton® PPS
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature CompensationYes
  • ATC ConnectorSpade lugs
  • ATC Element100 Ω Pt RTD
  • ConnectorBNC
  • DescriptionSolution Grounded pH Probe, DJ/PPS/100Ohm RTD; Plug/BNC
  • Warranty1 year
1 Year WarrantyKeep a Spare
Key Features
  • Get longer life with PTFE double junctions
  • Ideal for harsh conditions
  • 1 year warranty
More About this Item
These electrodes offer solution ground, PTFE references with double junctions and polymerized gel. All models include a 10-ft cable with BNC pH connector and Ryton® housing with 3/4'' NPT(M) threads.
General-Purpose Electrodes have low-resistant glass with a very fast and stable response. Designed for minimal salt applications. Use for measurements from 0 to 13 pH. Temperature range is 32 to 212°F (0 to 100°C).