Cole-Parmer Spex® 2201 Sealing Mat for Square 96-Well Microplate; 1/EA
Representative image only.
Cole-Parmer Item # EW-04575-65
Tightly seals microplate wells to prevents spills and well-to-well contamination
  • Constructed of rugged silicone rubber
  • May be sterilized and reused
  • Use with 96-well microplates with square wells (04575-64 and 04576-90)
  • 1 each
PRODUCT OPTIONS
$44.10USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialSilicone
  • Compatible WithMicroplates 04575-64 and 04576-90
  • DescriptionSealing Mat for Square 96-Well Microplate; 1/EA
Key Features
  • Constructed of rugged silicone rubber
  • May be sterilized and reused
  • Use with 96-well microplates with square wells (04575-64 and 04576-90)
  • 1 each
More About this Item
Use these flexible deep-well sealing mats to reduce maximum well volume. 