Cole-Parmer Spex® 2300 Homogenizer Foam Holder, 48 x 2 mL Tubes; 1/EA

Representative image only.
Cole-Parmer Item # EW-04500-23
Securely hold vials in place while processing samples
  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Thick and rugged, holds 48 microcentrifuge tubes

PRODUCT OPTIONS

$70.40USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Max Tube Size (mL)2
  • Tubes Held48
  • MaterialFoam
  • DescriptionHomogenizer Foam Holder, 48 x 2 mL Tubes; 1/EA

Key Features

  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Thick and rugged, holds 48 microcentrifuge tubes

More About this Item

Vial holders are suitable for use with the HG-600 Geno/Grinder® 2010 Homogenizer. Order homogenizer and vials separately.

Thick, rugged foam block holds 48 (2 mL) microfuge tubes.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat