Cole-Parmer Spex® 3642-450 Cellulose Binder Powder, <20 µm, 450 g
Representative image only.
Cole-Parmer Item # EW-04577-49
Prepare samples for reproducible and contamination-free results
  • Select binders from powder, tablet, or liquid form depending on your application
  • Cellulose powder with a particle size of <20 µm
  • 450 g bottle
  • Will bond 600 to 750 sample disks or provide matrix for 75 to 105 supported sample disks
PRODUCT OPTIONS
$124.00USD / Each
Don't forget to order scoops
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPowder
  • DescriptionCellulose Binder Powder, <20 µm, 450 g
Key Features
  • Select binders from powder, tablet, or liquid form depending on your application
  • Cellulose powder with a particle size of <20 µm
  • 450 g bottle
  • Will bond 600 to 750 sample disks or provide matrix for 75 to 105 supported sample disks
More About this Item
High-quality sample preparation is the key to achieving consistent and reliable analytical results. Powders are ideal for samples that crumble or ablate after pressing. They blend quickly and completely with samples to form XRF (x-ray fluorescence) sample disks. Tablets are an efficient way to add binder to XRF sample disks. Liquid easily mixes with the sample.
Form XRF sample disks by blending with sample at 10 to 25% by weight or use undiluted as sample matrix with sleeve-and-plunger set 04577-38 or 04577-45. Powder will not stick to the pellet die nor contaminate the sample.