Cole-Parmer Spex® 3646-450 Paraffin Binder Powder, <30 µm, 450 g
Representative image only.
Cole-Parmer Item # EW-04577-55
Prepare samples for reproducible and contamination-free results
  • Select binders from powder, tablet, or liquid form depending on your application
  • Paraffin powder with a particle size of <30 µm
  • 450 g bottle
  • Will bond 600 to 750 sample disks
PRODUCT OPTIONS
$101.00USD / Each
Don't forget to order scoops
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPowder
  • DescriptionParaffin Binder Powder, <30 µm, 450 g
Key Features
  • Select binders from powder, tablet, or liquid form depending on your application
  • Paraffin powder with a particle size of <30 µm
  • 450 g bottle
  • Will bond 600 to 750 sample disks
More About this Item
High-quality sample preparation is the key to achieving consistent and reliable analytical results. Powders are ideal for samples that crumble or ablate after pressing. They blend quickly and completely with samples to form XRF (x-ray fluorescence) sample disks. Tablets are an efficient way to add binder to XRF sample disks. Liquid easily mixes with the sample.
Air-stable disks act as the waxy binder to seal the surface against moisture. Paraffin binder can be blended with sample at 10 to 20% by weight to form XRF sample disks.