Cole-Parmer Spex® 51793 O-Ring Drive Belt for SM-200 ShatterBox; 1/EA
Representative image only.
Cole-Parmer Item # EW-04576-83
Optimize performance of your Shatterbox® Verastile Ring and Puck Mill
  • Choose from a wide range of accessories and replacement parts to ensure safe operation
  • Always replace as a set of four to ensure smooth, balanced operation
PRODUCT OPTIONS
$24.20USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialElastomer
  • Compatible WithShatterBox® Ring and Puck Mill
  • DescriptionO-Ring Drive Belt for SM-200 ShatterBox; 1/EA
Key Features
  • Choose from a wide range of accessories and replacement parts to ensure safe operation
  • Always replace as a set of four to ensure smooth, balanced operation
More About this Item
Suitable for use with the Cole-Parmer® SM-200 ShatterBox® Ring and Puck Mills, 04578-25 and 04578-26. Mills sold separately.
Elastomer drive belts link motor and grinding mechanism.