Cole-Parmer Spex® 6881C4 Replacement Cylinders for Mid-Size Grinding Vial Sets, PC; 4/PK
Representative image only.
Cole-Parmer Item # EW-04577-96
Keep spare items to ensure there is no down time
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Use with mid-size grinding vial sets 61043-08, 61043-12, and 61043-13
  • Clear polycarbonate construction
  • Pack of 4
PRODUCT OPTIONS
$213.25USD / Pkg of 4
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolycarbonate
  • Compatible WithMid-size grinding vial sets 61043-08, -12, and -13
  • DescriptionReplacement Cylinders for Mid-Size Grinding Vial Sets, PC; 4/PK
Key Features
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Use with mid-size grinding vial sets 61043-08, 61043-12, and 61043-13
  • Clear polycarbonate construction
  • Pack of 4
More About this Item
Optimize performance of your Cole-Parmer® Freezer/Mill® grinding vial sets with a wide range of grinding ball options suitable for your application.