Cole-Parmer Spex® 6885P Replacement Impactor for Mid-Size Grinding Vial Set 61043-13, PC Encapsulated Steel; 1/EA
Representative image only.
Cole-Parmer Item # EW-04578-02
Keep spare items to ensure there is no down time
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Polycarbonate encapsulated steel impactor
  • Eliminate metal contamination
  • Ideal for grinding soft plant and animal tissues
PRODUCT OPTIONS
$141.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPC encapsulated steel
  • Compatible WithMid-size grinding vial set 61043-13
  • DescriptionReplacement Impactor for Mid-Size Grinding Vial Set 61043-13, PC Encapsulated Steel; 1/EA
Key Features
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Polycarbonate encapsulated steel impactor
  • Eliminate metal contamination
  • Ideal for grinding soft plant and animal tissues
More About this Item
Optimize performance of your Cole-Parmer® Freezer/Mill® grinding vial sets with a wide range of grinding ball options suitable for your application.