Cole-Parmer Spex® 6888 Vial Holder Adapter for Mid-Size Vial, Closed-End
Representative image only.
Cole-Parmer Item # EW-04500-52
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Adapter for mid-size vial set 61043-13
  • Prevents sample leakage
PRODUCT OPTIONS
$500.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)25
  • Tubes Held1
  • Capacity1 mid-size vial
  • DescriptionVial Holder Adapter for Mid-Size Vial, Closed-End
Key Features
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Adapter for mid-size vial set 61043-13
  • Prevents sample leakage
More About this Item
Compatible with Freezer/Mill® cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.
Use this holder with mid-size grinding vial sets 61043-08 and 61043-12. It is open at the back end to allow the force of the impact to strike against the back anvil of the cryogenic grinder coil. Compatible with CG-400, CG-450, and CG-500 Freezer/Mill® cryogenic grinders.