Cole-Parmer Spex® 8221B-20 Hardened Steel Ball Set, 20mm; 20/Pk
Representative image only.
Cole-Parmer Item # EW-04900-08
Accessories to rapidly reduce hard and brittle samples with the BM-600 Planetary Mill
  • Jar rotation in combination with grinding balls impacting the sample and the walls of the jar allow for both milling and mixing
  • Excellent for grinding rocks, minerals, sand, cement, slag, ceramics, glass, and pharmaceuticals
  • 500 mL jar can hold 20 x 20 mm grinding balls and the 250 mL jar can hold 10 x 20 mm grinding balls
PRODUCT OPTIONS
$141.75USD / Pkg of 20
SPECS & DESCRIPTION
Specifications & Description
  • MaterialHardened Steel
  • Diameter (cm)2
  • Compatible With250 and 500 mL Grinding Jar
  • DescriptionHardened Steel Ball Set, 20mm; 20/Pk
Key Features
  • Jar rotation in combination with grinding balls impacting the sample and the walls of the jar allow for both milling and mixing
  • Excellent for grinding rocks, minerals, sand, cement, slag, ceramics, glass, and pharmaceuticals
  • 500 mL jar can hold 20 x 20 mm grinding balls and the 250 mL jar can hold 10 x 20 mm grinding balls
More About this Item
These accessories are compatible with the Cole-Parmer® BM-600 Planetary Mills 04900-16 and 04900-17. Mill sold separately.
Choose from 250 mL or 500 mL jar capacities and stainless steel or hardened steel jars and grinding media. Generally, the grinding jar and grinding balls are made from the same material.