Cole-Parmer Spex® 8511-05 Handle Assembly for Alumina Ceramic and Zirconia Grinding Containers
Representative image only.
Cole-Parmer Item # EW-41010-90
Easily convert your existing dish without handles
  • Optimize performance of your SM-300 ShatterBox®
  • Compatible with alumina ceramic (41010-89) and zirconia (41010-91) Grinding Containers
PRODUCT OPTIONS
$419.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithAlumina ceramic (04576-77) & zirocnia (04576-78) containers
  • DescriptionHandle Assembly for Alumina Ceramic and Zirconia Grinding Containers
Made in the U.S.A.
Key Features
  • Optimize performance of your SM-300 ShatterBox®
  • Compatible with alumina ceramic (41010-89) and zirconia (41010-91) Grinding Containers
More About this Item
The SM-300 ShatterBox® requires the use of a dish with a handle assembly. For existing dishes without handles, the corresponding handle assembly may be purchased and secured to the dish.