Cole-Parmer Spigot Fitting, 1 1/8" Female, 3/4" Hose Barb
Representative image only.
Cole-Parmer Item # EW-06061-66
Use these fittings and spigot with carboys to easily dispense liquid
  • Made of autoclavable polypropylene
  • Feature a 1-1/8"-12 UNF-2B thread
  • 3/4" hose barb provides a quick connection to tubing
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Sterilization MethodAutoclavable
  • Connection Type3/4\" hose barb
  • ColorWhite
  • DescriptionCarboy fitting, 1/pk
Key Features
  • Made of autoclavable polypropylene
  • Feature a 1-1/8"-12 UNF-2B thread
  • 3/4" hose barb provides a quick connection to tubing
More About this Item
Fittings are available in 1/4", 3/8", 1/2", and 3/4" hose barb sizes for convenient connection to tubing. Use the 3/4" sanitary connection fitting to make carboys function as a reservoir to a larger system. Spigot has a hose barb to accept 5/8" to 3/4" ID tubing and a standard internal luer taper to fit a male barb adapter. The internal thread has a hard stop to prevent over-tightening which may damage the seal. Use the large lever to control two flow rates—continuous locked flow or faster manual flow.