Cole-Parmer StableTemp 949138 Dry Block Heater Cover for 2-Block Models
Representative image only.
Cole-Parmer StableTemp Item # EW-36600-65
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Height (in)1 5/8
  • Width (in)6 1/2
  • Length (in)6 1/2
  • DescriptionDry Block Heater Cover for 2-Block Models
  • Warranty2 Years
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Choose from our comprehensive line of Cole-Parmer StableTemp Block Heaters Low Temperature Covers and other Cole-Parmer StableTemp Dry Block Heaters and Blocks products. Polycarbonate cover is designed to reduce the air flow for additional temperature stability in low-temperature applications. Two sides are open
Low temperature cover reduces air flow for added low temperature stability in low temperature applications. Fits 2 block models