Cole-Parmer StableTemp Block for Vials, 23mm Vials

Representative image only.
Cole-Parmer StableTemp Item # EW-36600-47
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Block Number Of Openings8
  • Block - Tube / Vial Size Accepted (mm)23
  • Height (in)2
  • Width (in)3
  • Length (in)3 3/4
  • DescriptionBlock for Vials, 23mm Vials
  • Warranty2 Years
Two year warranty

Key Features

  • 2 year warranty

More About this Item

Modular blocks are constructed from a solid anodized aluminum block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature.
Vial Blocks are single blocks and are designed for sample/serum and scintillation vials. Blocks are available for vial sizes from 12mm to 28mm in diameter.

Vial block for 23mm vials. Single block

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat