Cole-Parmer StableTemp Dry Block Heaters (Analog), 4 Block, 120V

Representative image only.
Cole-Parmer StableTemp Item # EW-36600-18
Precision temperature stability ensures reproducibility of results
  • Interchangeable modular heating blocks provide flexibility for heating different-size tubes -order blocks separately
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Number Of Blocks4
  • Uniformity (° C)±0.4°
  • Max Temperature (° F)302
  • Max Temperature (° C)150
  • Power (VAC)120
  • Power (watts)310
  • Height (in)3 1/2
  • Width (in)8
  • Length (in)16 29/32
  • DescriptionDry Block Heaters (Analog), 4 Block, 120V
  • Warranty2 Years
Two year warrantyConformite Europeenne

Key Features

  • Interchangeable modular heating blocks provide flexibility for heating different-size tubes -order blocks separately
  • 2 year warranty

More About this Item

Designed for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a wide variety of other laboratory procedures. Each heater accepts separate interchangeable modular heating blocks, which accommodate various sample enclosures such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes. Create a versatile system unique to your application; order either an analog or digital heater and the required heating block module (sold separately online).

CAUTION! To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed to run as a dry bath / incubator.

Analog Dry Block Heaters are economical, high-wattage, constant temperature heaters that are ideal for a wide variety of laboratory applications. They are controlled by two separate temperature adjustment knobs, with 1 to 10 dial markings for low and high temperature ranges. Low-range knob adjusts from ambient to 100ºC and high-range knob adjusts from 75 to 150ºC. A separate thermometer or probe is needed to set and verify the setpoint temperature.

Analog dry block heater 4 block capacity 120V

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat