Cole-Parmer StableTemp Dry Block Heaters External Temperature Probe Kit
Representative image only.
Cole-Parmer StableTemp Item # EW-36600-68
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDry Block Heaters External Temperature Probe Kit
  • Warranty2 Years
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Enables the digital unit to read the actual block or sample temperature and display that temperature on the control panel. Kit includes a PT-100 RTD temperature probe, 18" (45.7cm) stainless steel support rod, thermometer/probe clamp, and a hook connector. Probe tip is designed to fit in the thermometer well of each block.