Cole-Parmer StableTemp Single Block - Solid
Representative image only.
Cole-Parmer StableTemp Item # EW-36600-55
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Height (in)2
  • Width (in)3
  • Length (in)3 3/4
  • DescriptionSingle Block - Solid
  • Warranty2 Years
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Modular blocks are constructed from a solid anodized aluminum block. Solid Blocks are available in either a single or double block size and can drilled to a custom specification, used for drying slides, or as a low temperature hotplate.