Cole-Parmer Stainless steel utiltiy tray, 25"L x 15"W

Representative image only.
Cole-Parmer Item # EW-07277-23

$226.00USD / Each

BUY MORE & SAVE

  • 1-5$226.00USD / Each
  • 6-29$203.40USD / Each
  • 30-119$192.10USD / Each
  • 120+$180.80USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • MaterialStainless Steel
  • Overall Length (in)25
  • Overall Length (cm)63.5
  • Overall Width (in)15
  • Overall Width (cm)38.1
  • Overall Height (in)7/8
  • Overall Height (cm)2.2
  • DescriptionStainless steel utiltiy tray, 25"L x 15"W

More About this Item

Stainless steel utility trays serve multiple purposes in the laboratory. These heavy-gauge 304 stainless steel trays can withstand temperatures up to 650°F (343°C). Keep several on hand to transport supplies, instruments, and other items.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat