Cole-Parmer Essentials Pestle for Cell Strainer, Sterile, Polypropylene, Individually Wrapped, 5.5" L; 50/Cs
Representative image only.
Cole-Parmer Essentials Item # EW-01959-18
Better manipulate material in cell strainer
  • Pestle design improves manipulation of material in the strainer during use
  • Sterilized by gamma irradiation
  • DNase-& RNase-free
  • Non-pyrogenic
  • Convex pestle head with textured surface helps manipulate material in the cell strainer
  • 5.5" L for convenient hand manipulation
$60.70USD / Case of 50
Often Bought With
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • ColorGreen
  • Compatible WithAll Cell Strainers
  • Length (in)5.5
  • Length (cm)14
  • DescriptionPestle for Cell Strainer, Sterile, Polypropylene, Individually Wrapped, 5.5\" L; 50/Cs
Sterile
Key Features
  • Pestle design improves manipulation of material in the strainer during use
  • Sterilized by gamma irradiation
  • DNase-& RNase-free
  • Non-pyrogenic
  • Convex pestle head with textured surface helps manipulate material in the cell strainer
  • 5.5" L for convenient hand manipulation
More About this Item
The Cole-Parmer® Essentials pestle features a convex head with molded textured surface to better direct slippery material in the cell strainer. The 5.5” length allows for convenient and ergonomic hand manipulation, providing a faster, easier cell straining process. Pestles are supplied sterile (gamma irradiation) and come individually wrapped.
Pestles are supplied sterile (gamma irradiation) and come individually wrapped.