Cole-Parmer Sterile PETG Square Media Bottle,125 mL; 144/Cs

Representative image only.
Cole-Parmer Item # EW-06542-14
Leakproof, shatterproof, optically clear, and gamma sterilized
  • PETG is a safe alternative to glass—shatterproof
  • Secure your samples with leakproof seals (included)
  • Molded-in graduations—ideal for reference filling
  • Gamma irradiated for SAL 10-6 sterility
  • 24 bottles per pack, 6 packs per case; 144 bottles total

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)125
  • Capacity (oz)4
  • MaterialPolyethylene Terephthalate (PET)
  • Min Graduation (mL)25
  • Max Graduation (mL)125
  • Graduation Interval (mL)25
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size38/430
  • Cap ColorBlue
  • Bottle ColorTransparent
  • Overall Height (in)4 3/8
  • Overall Height (cm)11.2
  • Overall Depth (in)2 1/8
  • Overall Depth (cm)5.4
  • Overall Width (in)2 1/8
  • Overall Width (cm)5.4
  • DescriptionCole-Parmer Sterile PETG Square Media Bottle,125 mL; 144/Cs
Sterile

Key Features

  • PETG is a safe alternative to glass—shatterproof
  • Secure your samples with leakproof seals (included)
  • Molded-in graduations—ideal for reference filling
  • Gamma irradiated for SAL 10-6 sterility
  • 24 bottles per pack, 6 packs per case; 144 bottles total

More About this Item

Store buffers, culture media, and other critical fluids in these sterile bottles. Square shape saves precious shelf space; bottles can be placed closer together.

Bottles come in case quantities, and smaller packs within. Each case is specially packaged to maintain sterility—multi-barrier packaging.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat