Cole-Parmer Stopcock with Luer Connections; 1-way; male lock, Non-sterile
Representative image only.
Cole-Parmer Item # EW-30600-00
  • HDPE handle rotates 360 degrees.
PRODUCT OPTIONS
$35.05USD / Pkg of 10
BUY MORE & SAVE
  • 1-9$35.05USD / Pkg of 10
  • 10-49$31.55USD / Pkg of 10
  • 50-99$29.79USD / Pkg of 10
  • 100+$28.04USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Outlet PortMale Luer Lock
  • Inlet PortFemale Luer
  • Wetted MaterialsPolycarbonate
  • Flow PatternOne-Way
  • DescriptionStopcock with Luer Connections; 1-way; male lock, Non-sterile
Key Features
  • HDPE handle rotates 360 degrees.
More About this Item
Choose stopcocks with either a male luer lock or a male luer slip. Handle rotates 90deg on one-way stopcocks, 180deg on three-way stopcocks, and 360deg on four-way stopcocks. All have PC body and HDPE stem.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
REVIEWS
REVIEW SNAPSHOT®
by PowerReviews
Cole-Parmer Stopcock with Luer Connections; 1-way; male lock, Non-sterile
2.7
(based on 6 reviews)
33%
of respondents would recommend this to a friend.
Pros
  • Easy to use (5)
Cons
  • Not durable (4)
Best Uses
No Best Uses
    • Reviewer Profile:
    • Academic/professional researcher (3)
Reviewed by 6 customers
Displaying reviews 1-6
Back to top
2.0
Not as pictured; these crack in months
By 
from San Antonio, TX
About Me Academic/Professional Researcher
Verified Reviewer
Pros
  • Easy To Use
Cons
  • Breaks Easily
Best Uses
  • In The Lab
Comments about Cole-Parmer Stopcock with Luer Connections; 1-way; male lock, Non-sterile:
(These are not as pictured but have been replaced by an inferior product!) These crack and then leak fluid after 1 to several months of normal, easy use and have to be replaced. They crack at the juncture with the turn-valve, but these valves have to be turned to use the product properly, of course. We use typical physiological salines at room temperature, nothing harsh. As in another review here, we were looking for the blue-handled, clear plastic ones because those are sturdy and withstand several years of easy use. Instead, these are white-handled, purplish-clear plastic like the ones we were trying to replace because of cracking and subsequent leakage.
2.0
Not as pictured; these crack in months
By 
from San Antonio, TX
About Me Academic/Professional Researcher
Pros
  • Easy To Use
Cons
  • Breaks Easily
Best Uses
    Comments about Cole-Parmer Stopcock with Luer Connections; 1-way; male lock, Non-sterile:
    (These are not as pictured but have been replaced by an inferior product!) These crack and then leak fluid after 1 to several months of normal, easy use and have to be replaced. They crack at the juncture with the turn-valve, but these valves have to be turned to use the product properly, of course. We use typical physiological salines at room temperature, nothing harsh. As in another review here, we were looking for the blue-handled, clear plastic ones because those are sturdy and withstand several years of easy use. Instead, these are white-handled, purplish-clear plastic like the ones we were trying to replace because of cracking and subsequent leakage.
    (1 of 1 customers found this review helpful)
    2.0
    No rotation lock
    By 
    from MA
    Verified Reviewer
    Comments about Cole-Parmer Stopcock with Luer Connections; 1-way; male lock, Non-sterile:
    Not as pictured. Does not have rotation lock to stop at 90 degrees as advertised. The version of this item previously supplied did (the blue handled, branded version), the current (white handled, generic) does not. New version leakier, seems ot be generally inferior product.
    5.0
    Good
    By 
    from Baton Rouge, LA
    About Me Academic/Professional Researcher
    Verified Reviewer
    Pros
    • Accurate
    • Clear Markings
    • Easy To Use
    • Sturdy
    Cons
      Best Uses
        Comments about Cole-Parmer Stopcock with Luer Connections; 1-way; male lock, Non-sterile:
        we are using these stopcocks for regular air samples. They are sturdy and works fine for our purposes.
        2.0
        Not reliable for frequent use
        By 
        from Cleveland, OH
        About Me Lab Assistant
        Verified Buyer
        Pros
        • Easy To Use
        Cons
        • Breaks Easily
        Best Uses
        • In The Lab
        • Research
        Comments about Cole-Parmer Stopcock with Luer Connections; 1-way; male lock, Non-sterile:
        This stopcock is easy to use, but often develops hairline cracks after turning the knob frequently during use. The hairline cracks are large enough to allow fluid to drip out. If working with a critical material, I would recommend using the PVDF or stainless steel versions of this stopcock instead of this polycarbonate version.
        (5 of 5 customers found this review helpful)
        3.0
        Usable
        By 
        from kansas
        About Me Lab Assistant
        Verified Buyer
        Pros
        • Easy To Use
        Cons
        • Breaks Easily
        • Markings Are Hard To See
        Best Uses
        • Research
        Comments about Cole-Parmer Stopcock with Luer Connections; 1-way; male lock, Non-sterile:
        Useful and gets the job done. Would not Does not allow air to slip by but can be tough to get open and allow the right amount of air out if you are stuck for time.
        Displaying reviews 1-6
        Back to top
        Q&A
        Do you have questions about this product?
        Get answers from real customers and in-house experts with AnswerBox.
        5 Questions | 6 Answers
        Displaying questions 1-5
        • Q:
          what is the maximum pressure that the stopcock can resist?
          Asked on 6/12/2012 by Anonymous
          2 answers
          • STAFF EXPERT
            A:
            I will email you with an answer to this question.
            Answered on 6/14/2012 by Jay from Customer Service
          • STAFF EXPERT
            A:
            ISO requires a 45 psi pressure and this unit will handle at least this amount. The unit may support higher pressures, but these units are not recommended for use above 45psi.
            Answered on 6/20/2012 by Jay from Customer Service
        • Q:
          is it screw thread interface?Can it match with the 60ml screw thread syringe?
          Asked on 5/30/2012 by Jing from China
          1 answer
          • Staff Reviewer
            A:
            Luer connections are standardized. Most syringes also have luer connections, regardless of the volume capacity, so the answer is most likely, yes this should connect directly.
            Answered on 6/4/2012 by Jen from Technical Support
        • Q:
          How can this be sterilised?
          Asked on 3/22/2011 by Kimbo78140 from Paris, France
          1 answer
          • Staff Reviewer
            A:
            These can be gamma sterilized. These are not autoclavable.
            Answered on 3/23/2011 by Jen from Technical Support
        • Q:
          is this product sterile?
          Asked on 1/28/2011 by Anonymous
          1 answer
          • Staff Reviewer
            A:
            No, this product is not sterile.
            Answered on 1/31/2011 by Jen from Technical Support
        • Q:
          What are the dimensions for this? What size opening will it fit?
          Asked on 6/21/2010 by Anonymous
          1 answer
          • Staff Reviewer
            A:
            The bore is 0.115" through the stopcock. The total length is roughly 2 inches. These have standard male slip tip and female luer lock connections. Please contact our technical application specialists for additional assistance (contact information can be found in the "contact us" section on the web page).
            Answered on 6/23/2010 by Jen from Technical Support
        Displaying questions 1-5
        Do you have a question about this product? 