Cole-Parmer Stopcocks with Luer Connections, 3-way, Male Lock, Non-sterile; 10/Pk
Representative image only.
Cole-Parmer Item # EW-30600-02
  • 3-way stopcock/flow diverter with male luer lock connections
  • Pack of 10
PRODUCT OPTIONS
$30.05USD / Pkg of 10
BUY MORE & SAVE
  • 1-9$30.05USD / Pkg of 10
  • 10-49$27.05USD / Pkg of 10
  • 50-99$25.54USD / Pkg of 10
  • 100+$24.04USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Outlet PortMale Luer Lock
  • Inlet PortFemale Luer
  • Wetted MaterialsPolycarbonate
  • Flow PatternThree-Way
  • DescriptionStopcocks with Luer Connections; 3-way; male lock, Non-sterile
Key Features
  • 3-way stopcock/flow diverter with male luer lock connections
  • Pack of 10
More About this Item
Choose stopcocks with either a male luer lock or a male luer slip. Handle rotates 90deg on one-way stopcocks, 180deg on three-way stopcocks, and 360deg on four-way stopcocks. All have PC body and HDPE stem.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
REVIEWS
REVIEW SNAPSHOT®
by PowerReviews
Cole-Parmer Stopcocks with Luer Connections; 3-way; male lock, Non-sterile
3.4
(based on 7 reviews)
57%
of respondents would recommend this to a friend.
Pros
  • Easy to use (4)
  • Accurate (3)
  • Clear markings (3)
  • Durable (3)
Cons
No Cons
Best Uses
No Best Uses
    • Reviewer Profile:
    • Academic/professional researcher (3)
Reviewed by 7 customers
Displaying reviews 1-7
Back to top
5.0
Simple, effective
By 
from Southampton UK
Verified Buyer
Comments about Cole-Parmer Stopcocks with Luer Connections; 3-way; male lock, Non-sterile:
As described. Does its job.
5.0
Good
By 
from Sacramento, CA
Verified Buyer
Comments about Cole-Parmer Stopcocks with Luer Connections; 3-way; male lock, Non-sterile:
As advertised
1.0
I would not recommend this product.
By 
from Canada
Verified Reviewer
Pros
    Cons
    • Crack during use
    • Leak liquid
    Best Uses
      Comments about Cole-Parmer Stopcocks with Luer Connections; 3-way; male lock, Non-sterile:
      These valves crack and leak liquid when attaching tubing and/or turning the valve during use. Since they do not look like the valve in the photo (they are actually made of clear purple plastic and have white valves), I believe that this product has been replaced by one made of inferior material (see previous post by CS).
      (1 of 1 customers found this review helpful)
      2.0
      Not as pictured; these crack in months
      By 
      from San Antonio, TX
      About Me Academic/Professional Researcher
      Verified Reviewer
      Pros
      • Easy To Use
      Cons
      • Breaks Easily
      Best Uses
      • In The Lab
      Comments about Cole-Parmer Stopcocks with Luer Connections; 3-way; male lock, Non-sterile:
      (These are not as pictured but have been replaced by an inferior product!) These crack and then leak fluid after 1 to several months of normal, easy use and have to be replaced. They crack at the juncture with the turn-valve, but these valves have to be turned to use the product properly, of course. We use typical physiological salines at room temperature, nothing harsh. As in another review here, we were looking for the blue-handled, clear plastic ones because those are sturdy and withstand several years of easy use. Instead, these are white-handled, purplish-clear plastic like the ones we were trying to replace because of cracking and subsequent leakage.
      (2 of 2 customers found this review helpful)
      5.0
      just right
      By 
      from Florence
      About Me Lab Assistant
      Pros
      • Accurate
      • Clear Markings
      • Easy To Use
      • Sturdy
      Cons
        Best Uses
        • Research
        Comments about Cole-Parmer Stopcocks with Luer Connections; 3-way; male lock, Non-sterile:
        great item
        (2 of 2 customers found this review helpful)
        3.0
        good product
        By 
        from KC KS
        About Me Academic/Professional Researcher
        Pros
        • Accurate
        • Clear Markings
        • Durable
        • Easy To Use
        • Sturdy
        Cons
        • Female does not rotate
        Best Uses
        • Imaging chamber perfusion
        • In The Lab
        • Research
        Comments about Cole-Parmer Stopcocks with Luer Connections; 3-way; male lock, Non-sterile:
        Would be a much better product if the female end was able to rotate.
        (2 of 2 customers found this review helpful)
        3.0
        As good as the materials
        By 
        from Flagstaff, AZ
        About Me Academic/Professional Researcher
        Verified Reviewer
        Pros
        • Accurate
        • Clear Markings
        • Easy To Use
        Cons
        • Breaks Easily
        Best Uses
          Comments about Cole-Parmer Stopcocks with Luer Connections; 3-way; male lock, Non-sterile:
          I set up the stopcock to quickly switch between mobile phase lines for an LCMS system. Unfortunately, the organics pumped through the lines weakened the polycarbonate (PC) valve piece. I should have checked if the PC parts were compatible with the liquids. Next time I'll try the polyvinylidene fluoride (PVDF)and polypropylene (PP)stopcock.
          Displaying reviews 1-7
          Back to top
          Q&A
          Do you have questions about this product?
          Get answers from real customers and in-house experts with AnswerBox.
          27 Questions | 28 Answers
          Displaying questions 1-10Previous | Next »
          • Q:
            Can you supply a ROHS compliance certificate for this product?"
            Asked on 1/31/2017 by Mike from Oak Ridge TN
            1 answer
            • STAFF EXPERT
              A:
              A RoHS certificate is available after purchase.
              Answered on 2/16/2017 by Mike from Tech
          • Q:
            what is the chemical compatibility with sodium hydroxide, 1 Normal, or 10%?
            do you have a 4 way valve that has good compatibility with 10% Sodium Hydroxide? the weld is the weak point.
            Asked on 7/13/2016 by suuuuuu from boston, ma
            1 answer
            • STAFF EXPERT
              A:
              There is no data for the 10% NaOH solution, but for 25% NaOH, this stopcock is shows excellent compatibility (up until 48C).
              For a 4 way valve, the item 30600-04 has the same type of connections as the 30600-02. There are also 30600-04 & 30600-12, which have Luer fittings and large-bore stopcocks respectively.
              Answered on 8/1/2016 by Armen from Technical Support
          • Q:
            What is the life cycle for the 30600-02 valve
            Asked on 7/4/2016 by Luis from Costa Rica
            1 answer
            • STAFF EXPERT
              A:
              It is hard to determine how long this stopcock will last. It all depends on your application. Please contact us directly at techinfo@coleparmer.com for more information.
              Answered on 8/24/2016 by Dan C from Technical Support
          • Q:
            Could i connect to female luers of this stopcock man luers size 1/16 inch (GY-45518-00)?
            Asked on 5/16/2016 by Atida
            1 answer
            • STAFF EXPERT
              A:
              Yes, this stopcock has 2 female luer locks and a male luer lock. Therefore, you would need the corresponding male or female luer lock to attach to the ports.
              Answered on 5/16/2016 by Rishi from Technical Support
          • Q:
            Can I connect this to size 3/32 (inner diameter) tubing ? What adapter do I need?
            Asked on 5/12/2016 by Xitong from United States
            1 answer
            • STAFF EXPERT
              A:
              This stopcock has 2 female luer locks and a male luer lock. Therefore, you would need the corresponding male or female luer lock to attach to the ports. A female by 3/32" hose barb is 45501-02 and the male by 3/32" barb is 45504-02.
              Answered on 5/13/2016 by Danielle from Technical Support
          • Q:
            Can I connect this to size 16 tubing? What adapter do I need?
            Do you have autoclavable this kind of 3-way connector?
            Asked on 2/12/2016 by Geelsu
            1 answer
            • STAFF EXPERT
              A:
              You will require a 1/8" barb to luer adapter similar to the 45501-04.
              Answered on 2/12/2016 by Polo from Technical Support
          • Q:
            As well as giving an 'L' and 'I' flow pattern will it also switch all connectors off?
            Asked on 12/29/2015 by caddy-carazy from UK
            1 answer
            • STAFF EXPERT
              A:
              Only one branch can be closed at a time.
              Answered on 12/29/2015 by Polo from Technical Support
          • Q:
            Can be connected with High-Temp Silicon Rubber Tubing, Soft, 3/8"ID, 5/8" OD at the temp range from 0 Celsius to 130 Celsius.
            Asked on 10/26/2015 by Lucian from Andover, MA
            1 answer
            • STAFF EXPERT
              A:
              You cannot connect this stopcock directly to tubing without an adapter. The largest tubing recommend to connect to a luer fitting is 1/4" ID, which would require a 45501-20 adapter.
              Answered on 10/26/2015 by Danielle from Technical Support
          • Q:
            I need this exact piece but with a rotating male lock (That would allow for no rotation of the arms while connecting - no spinning)
            Asked on 8/8/2015 by Researcherrerrerr from Clemson, SC
            1 answer
            • STAFF EXPERT
              A:
              This is only available with a locking ring which does not rotate.
              Answered on 8/10/2015 by Dan from Technical Support
          • Q:
            Is this product autoclavable?
            Asked on 5/12/2015 by Sam from Hamilton, UK
            1 answer
            • STAFF EXPERT
              A:
              These are not autoclavable.
              Answered on 5/12/2015 by Polo from Technical Support
          Displaying questions 1-10Previous | Next »
          Do you have a question about this product? 