Cole-Parmer Sulfuric Acid, 1.00 Normal (1 gal) (Liquid)

Representative image only.
Cole-Parmer Item # EW-88401-08
Reagents and solutions to meet your lab and research needs

PRODUCT OPTIONS

$65.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Normality1
  • Purity GradeComponents all ACS
  • Linear FormulaH2SO4
  • ClassTitrants
  • ChemistryAcids;Inorganics
  • FormLiquid
  • SynonymsHygrogen sulfate; Oil of vitriol; Vitriol brown oil; Mattling acid; Battery acid; Sulphuric acid; Electrolyte acid; Dihydrogen sulfate; Spirt of sulphur; Chamber acid
  • Volume (mL)4000
  • Formula Weight98.08
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number7732-18-5, 7664-93-9
  • Shelf Life (months)24
  • DescriptionSulfuric Acid, 1.00 Normal (1 gal)

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat