Cole-Parmer Sulfuric Acid Solution, ACS Grade, Safety-Coated Glass Bottle, 2.5 L

Representative image only.
Cole-Parmer Item # EW-88406-06
Reagents and solutions to meet your lab and research needs
  • Quality and consistency your lab can count on

PRODUCT OPTIONS

$187.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration0.02
  • Purity GradeACS Reagent
  • Empirical FormulaH2SO4
  • ClassReagent Acids and Bases
  • ChemistryAcids
  • FormLiquid
  • SynonymsHygrogen sulfate, Dihydrogen sulfate
  • Volume (mL)2500
  • Formula Weight98.07
  • PackagingSafety Coated Glass Bottle
  • DOT InformationSulfuric acid; Class 8, Packing Group II
  • Storage SymbolWhite
  • Hazard SymbolCorrosive
  • CAS Number7664-93-9
  • UN NumberUN1830
  • Shelf Life (months)24
  • DescriptionSulfuric Acid Solution, ACS Grade, Safety-Coated Glass Bottle, 2.5 L

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Please note this product may require additional documentation/verification to purchase. This may include business identification and an authorized purchaser form:
Acceptable business identification
State issued document with your business information

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat