Cole-Parmer TELOS® Polyethylene Frits 20 µm, 150 mL

Representative image only.
Cole-Parmer Item # EW-06479-84
Optimize performance of your TELOS reservoirs
  • Available in 10 or 20 µm
  • 20 µm PE for 150 mL columns

PRODUCT OPTIONS

$77.50USD / Pkg of 50

SPECS & DESCRIPTION

Specifications & Description

  • Wetted MaterialsPolyethylene
  • Fritted Disk20 µm
  • DescriptionTELOS Polyethylene Frits 20 µm, 150 mL

Key Features

  • Available in 10 or 20 µm
  • 20 µm PE for 150 mL columns

More About this Item

Choose from multiple frit material options ideal for your application needs.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat