Cole-Parmer Temperature Compensating Probe for Ultrapure Water System, 100 Ohm PT RTD
Representative image only.
Cole-Parmer Item # EW-27004-22
Optimize performance of your Ultrapure Water System
  • Easily measure the temperature of your ultrapure water
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsUltrapure
  • Temperature CompensationYes
  • ATC ConnectorMini-audio plug
  • ATC Element100 Ω Pt RTD
  • Cable Length (m)3
  • DescriptionTemperature Compensating Probe for Ultrapure Water System, 100 Ohm PT RTD
  • Warranty1 year
1 Year Warranty
Key Features
  • Easily measure the temperature of your ultrapure water
  • 1 year warranty
More About this Item
Temperature probe (upper left), pH electrode (upper right), and flow cell (bottom) shown in image.
Temperature probe (upper left), pH electrode (upper right), and flow cell (bottom) shown in image.