Cole-Parmer Testing Sieve, 8" OD Brass Frame/Brass Wire, Full Height, No. 170

Representative image only.
Cole-Parmer Item # EW-59980-19
Get more accurate results
  • Sievers are serialized
  • Come with a test sieve certificate of compliance
  • 1 year warranty

PRODUCT OPTIONS

$103.00USD / Each

$157.00 - $230.00USD / Each

for an additional
$157.00USD / Each
for an additional
$230.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Cole-Parmer Air Duster Spray, 10 oz; 6/Pk
$76.50USD / Case of 6
$115.50USD / Case of 6

SPECS & DESCRIPTION

Specifications & Description

  • MaterialBrass frame, brass wire
  • U.S. Standard Screen Size (No.)170
  • Tyler Standard Screen Size (mesh)170
  • Sieve Opening (mm)0.088
  • Outside Diameter (in)8
  • Height (InchShort)2
  • DescriptionTesting Sieve, 8" OD Brass Frame/Brass Wire, Full height, No. 170
  • Warranty1 Year
1 Year WarrantyISO9001 Certified Supplier

Key Features

  • Sievers are serialized
  • Come with a test sieve certificate of compliance
  • 1 year warranty

More About this Item

These brass testing sieves are serialized and come with a test sieve certificate of compliance. Equivalent to ASTM E11-09 compliance sieve.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat