Cole-Parmer Essentials Testing Sieve, 8" SS Frame/SS Wire, Half Height, No. 25
Representative image only.
Cole-Parmer Essentials Item # EW-59985-07
Don't second guess accuracy
  • Come with test sieve certificate of compliance
  • Serialized
  • 1 year warranty
PRODUCT OPTIONS
$122.50USD / Each
$157.00 - $230.00USD / Each
for an additional
$157.00USD / Each
for an additional
$230.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • U.S. Standard Screen Size (No.)25
  • Tyler Standard Screen Size (mesh)24
  • Sieve Opening (mm)0.707
  • Outside Diameter (in)8
  • Height (InchShort)1
  • DescriptionTesting Sieve, 8\" SS Frame/SS Wire, half height, No. 25
  • Warranty1 Year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Come with test sieve certificate of compliance
  • Serialized
  • 1 year warranty
More About this Item
These stainless steel (SS) testing sieves are serialized and come with a test sieve certificate of compliance. Equivalent to ASTM E11-09 compliance sieve.