Cole-Parmer Essentials Testing Sieve, Brass Frame/SS Wire, 8" OD, Half Height, No. 45
Representative image only.
Cole-Parmer Essentials Item # EW-59941-11
  • Economical brass frame combined with the durability of stainless steel mesh
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialBrass frame, stainless steel wire
  • U.S. Standard Screen Size (No.)45
  • Tyler Standard Screen Size (mesh)42
  • Sieve Opening (mm)0.355
  • Outside Diameter (in)8
  • Height (InchShort)1
  • DescriptionTesting Sieve, Brass Frame/SS Wire, 8\" OD, half height, No. 45
  • Warranty1 Year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Economical brass frame combined with the durability of stainless steel mesh
  • 1 year warranty
More About this Item
These brass and stainless steel (SS) testing sieves are serialized and come with a test sieve certificate of compliance. Equivalent to ASTM E11-09 compliance sieve.