Cole-Parmer Essentials Testing Sieve, SS Frame/SS Wire, 12" OD, Half Height, No. 140
Representative image only.
Cole-Parmer Essentials Item # EW-59995-18
Get more accurate results
  • Sievers are serialized
  • Come with a test sieve certificate of compliance
  • 1 year warranty
PRODUCT OPTIONS
$260.75USD / Each
$157.00 - $230.00USD / Each
for an additional
$157.00USD / Each
for an additional
$230.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • U.S. Standard Screen Size (No.)140
  • Tyler Standard Screen Size (mesh)150
  • Sieve Opening (mm)0.105
  • Outside Diameter (in)12
  • Height (InchShort)1.625
  • DescriptionTesting Sieve, SS Frame/SS Wire, 12\" OD, Half height, No. 140
  • Warranty1 Year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Sievers are serialized
  • Come with a test sieve certificate of compliance
  • 1 year warranty
More About this Item
These stainless steel (SS) testing sieves are serialized and come with a test sieve certificate of compliance. Equivalent to ASTM E11-09 compliance sieve.