Cole-Parmer ThinTouch™ Nitrile Blue Gloves, 9.5, Medium 100/Box
Representative image only.
Cole-Parmer Item # EW-15954-43
Enhanced tactile sensitivity for a superior “touch & feel” experience
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Advanced nitrile formulation with soft finish delivers optimal comfort and dexterity
  • Reliable protection against a wide range of chemicals
  • Textured fingertips ensure a secure grip in wet or dry conditions
  • High tensile strength for excellent tear and puncture resistance
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)5.1
  • SizeMedium
  • Length (in)9 1/2
  • Length (cm)24
  • ColorBlue
  • GripTextured fingertips
  • CuffBeaded
  • SterileNo
  • PowderedNo
  • DescriptionThinTouch™ Nitrile Blue Gloves, 9.5, Medium 100/Box
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Advanced nitrile formulation with soft finish delivers optimal comfort and dexterity
  • Reliable protection against a wide range of chemicals
  • Textured fingertips ensure a secure grip in wet or dry conditions
  • High tensile strength for excellent tear and puncture resistance
More About this Item
These gloves are soft, latex-free and powder-free. Available in 9.5” length and four sizes – small, medium, large and x-large. Textured fingertips enhance wet or dry grip. Ambidextrous with beaded cuffs.
AQL: 1.5