Cole-Parmer Three Tray Gel Caster for Horizontal Mini-Gel System Model 28559-00
Representative image only.
Cole-Parmer Item # EW-28559-82
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionThree Tray Gel Caster for Horizontal Mini-Gel System Model 28559-00
  • Warranty3 year
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
Gel Casters allow you to precast flat, even gels using the gel trays sold above. Casters accept one, three, or six trays. Casters feature a three-point leveling base and soft neoprene side gaskets that secure trays in place and effectively seal their ends during casting.