Cole-Parmer Three Tube Frame for 150 mm Flowtube, Common, Aluminum
Representative image only.
Cole-Parmer Item # EW-03215-30
$279.00USD / Each
BUY MORE & SAVE
  • 1-3$279.00USD / Each
  • 4-9$251.10USD / Each
  • 10-19$237.15USD / Each
  • 20+$223.20USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Operating Temperature (° F)250
  • Max Operating Temperature (° C)121
  • Process Connection (NPT(F))0
  • DescriptionThree Tube Frame for 150 mm Flowtube, Common, Aluminum
More About this Item
These frames have valve ports which accept valve cartridges to control flow. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting.