Cole-Parmer Toluene, ACS Grade, 1 L

Representative image only.
Cole-Parmer Item # EW-88406-20
Reagents and solutions to meet your lab and research needs
  • Quality and consistency your lab can count on
  • Assay: min 99.5%
  • Color (APHA): max 10
  • Residue after evaporation (Non-Volatile Matter): max 0.001%
  • Substances Darkened by Sulfuric Acid: To Pass Test
  • Sulfur compounds (as S): max 0.003%
  • Water (H2O): max 0.03%

PRODUCT OPTIONS

$169.25USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • Empirical FormulaC6H5CH3
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsMethylbenzene, Toluol, Phenyl Methane
  • Volume (mL)1000
  • Formula Weight92.13
  • PackagingAmber Glass Bottle
  • DOT InformationToluene; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolFlammable, Harmful
  • CAS Number108-88-3
  • UN NumberUN1294
  • Shelf Life (months)12
  • DescriptionToluene, ACS Grade, 1 L
FlammableSolventStorage Code Red

Key Features

  • Quality and consistency your lab can count on
  • Assay: min 99.5%
  • Color (APHA): max 10
  • Residue after evaporation (Non-Volatile Matter): max 0.001%
  • Substances Darkened by Sulfuric Acid: To Pass Test
  • Sulfur compounds (as S): max 0.003%
  • Water (H2O): max 0.03%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Please note this product may require additional documentation/verification to purchase. This may include business identification and an authorized purchaser form:
Acceptable business identification
State issued document with your business information

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat