Cole-Parmer Triton® X-100 Wetting Agent, 1 L

Representative image only.
Cole-Parmer Item # EW-88401-18
Reagents and solutions to meet your lab and research needs
  • This nonionic detergent is used to solubilize membranes, permeabilize unfixed eukaryotic membranes, and is a component of lysis buffers for DNA extraction

PRODUCT OPTIONS

$117.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC14H22O(OCH2CH2)_n
  • ChemistrySolvents
  • FormLiquid
  • Volume (mL)1000
  • Formula Weight206.32
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number9002-93-1
  • Shelf Life (months)24
  • DescriptionTriton® X-100 Wetting Agent, 1 L

Key Features

  • This nonionic detergent is used to solubilize membranes, permeabilize unfixed eukaryotic membranes, and is a component of lysis buffers for DNA extraction

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat