Cole-Parmer Tuff-Tip, Semi-Dome 1" In-Line/Sub pH, graphite, 1kOhm spade lug ATC
Representative image only.
Cole-Parmer Item # EW-27020-66
Less chance of breakage
  • Choose chemical-resistant Ryton® PPS or graphite-tipped electrodes
PRODUCT OPTIONS
$346.00USD / Each
BUY MORE & SAVE
  • 1-5$346.00USD / Each
  • 6-11$311.40USD / Each
  • 12-23$294.10USD / Each
  • 24+$276.80USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode Body MaterialKynar®
  • Reference JunctionCeramic
  • Max Temperature (° C)110
  • Temperature CompensationYes
  • ATC ConnectorSpade lugs
  • ATC Element1 kΩ Pt RTD
  • ConnectorBNC
  • DescriptionTuff-Tip, Semi-Dome 1" In-Line/Sub pH, graphite, 1kOhm spade lug ATC
Key Features
  • Choose chemical-resistant Ryton® PPS or graphite-tipped electrodes
More About this Item
Don't worry about bulb breakage— unique design helps protect bulbs
– Eliminate electrical noise with graphite-tipped models
– Ideal for viscous or severe applications
– Reduce contamination and clogging with uniform pore size