Cole-Parmer Tuff-Tip, Semi-Domed 1" In-Line or Submersible 100 Ohm PT RTD, BNC
Representative image only.
Cole-Parmer Item # EW-27020-22
Less chance of breakage
  • Choose chemical-resistant Ryton® PPS or graphite-tipped electrodes
  • 1 year warranty
PRODUCT OPTIONS
$293.00USD / Each
BUY MORE & SAVE
  • 1-5$293.00USD / Each
  • 6-11$263.70USD / Each
  • 12-23$249.05USD / Each
  • 24+$234.40USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsIn-line or submersible
  • Electrode TypeDouble junction
  • Electrode Body MaterialKynar®
  • Reference JunctionCeramic
  • Max Temperature (° C)110
  • Temperature CompensationYes
  • ATC ConnectorTinned ends
  • ATC Element100 Ω Pt RTD
  • ConnectorBNC
  • DescriptionTuff-Tip, Semi-Domed 1\" In-Line or Submersible 100 Ohm PT RTD, BNC
  • Warranty1 year
1 Year WarrantyKeep a Spare
Key Features
  • Choose chemical-resistant Ryton® PPS or graphite-tipped electrodes
  • 1 year warranty
More About this Item
Don't worry about bulb breakage— unique design helps protect bulbs
– Eliminate electrical noise with graphite-tipped models
– Ideal for viscous or severe applications
– Reduce contamination and clogging with uniform pore size