Cole-Parmer Tween 80, 500 mL

Representative image only.
Cole-Parmer Item # EW-88401-58
Non-ionic detergent for selective protein extraction
  • Vapor Pressure: <1hPa at 20°C
  • Merck Index: 15, 7703
  • Ignition residue: 0.25% max
  • Density: 1.080 g/cm3

$253.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • ChemistrySolvents
  • FormLiquid
  • SynonymsPolyoxyethylene 20 Sorbitan Monooleate
  • Volume (mL)500
  • PackagingAmber Glass Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number9005-65-6
  • Shelf Life (months)12
  • DescriptionTween 80, 500 mL

Key Features

  • Vapor Pressure: <1hPa at 20°C
  • Merck Index: 15, 7703
  • Ignition residue: 0.25% max
  • Density: 1.080 g/cm3

More About this Item

This nonionic detergent is used in the isolation of water-soluble membrane proteins.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat