Cole-Parmer Essentials Twistit white rubber stopper, size 7
Representative image only.
Cole-Parmer Essentials Item # EW-62992-26
$72.80USD / Pkg of 20
BUY MORE & SAVE
  • 1-5$72.80USD / Pkg of 20
  • 6-29$65.52USD / Pkg of 20
  • 30-119$61.88USD / Pkg of 20
  • 120+$58.24USD / Pkg of 20
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number7
  • Top Outside Diameter (OD) (mm)37
  • Bottom Outside Diameter (OD) (mm)30
  • Height (mm)25
  • ColorWhite
  • MaterialRubber
  • DescriptionTwistit white rubber stopper, size 7
More About this Item
Use these versatile Twistit stoppers as either a solid, one-hole, two-hole, or three-hole stopper. Simply twist off a bottom seal to open a hole. The soft resilient natural rubber stays pliable over a long period and withstands laboratory conditions without hardening. Stoppers retain resiliency even after repeated autoclaving. Each stopper has its size in raised figures on top. Available in black or white rubber. Autoclavable.