Cole-Parmer Essentials Two-hole Rubber Stopper Size 6 1/2 21/pk
Representative image only.
Cole-Parmer Item # EW-62991-78
$84.50USD / Pkg of 21
BUY MORE & SAVE
  • 1-5$84.50USD / Pkg of 21
  • 6-29$76.05USD / Pkg of 21
  • 30-119$71.83USD / Pkg of 21
  • 120+$67.60USD / Pkg of 21
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number6.5
  • Top Outside Diameter (OD) (mm)34
  • Bottom Outside Diameter (OD) (mm)27
  • Height (mm)25
  • ColorBlack
  • MaterialRubber
  • DescriptionTwo-hole Rubber Stopper Size 6 1/2 21/pk
More About this Item
These two-hole black stoppers made with solid natural rubber stay pliable over a long period of time. Provide good tightness and adhesion, withstand laboratory conditions without hardening, and are ideal for use with nonaggressive or low concentration solutions. Each stopper has its size marked in raised figures. Low sulfur content. Withstand temperatures from –13 to 158°F (–25 to 70°C).