Cole-Parmer Two-way normally open solenoid pinch valve; 24 VDC, 1/8" ID x 1/4" OD tubing
Representative image only.
Cole-Parmer Item # EW-98302-36
No contamination of valve
  • Control flow in your tubing without direct contact between the fluid and the valve
  • Fast response time
  • Low-power consumption and high-cycle life
  • Excellent for sanitary and pharmaceutical applications
PRODUCT OPTIONS
$166.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Pinch Valve Tube Size1/8" ID x 1/4" OD
  • Connection TypePinch Valve for Tubing
  • Max Pressure (PSI)20
  • Power (VDC)24
  • Watts4
  • Response35 msec
  • Flow PatternTwo-Way, Normally Closed
  • DescriptionTwo-way normally open solenoid pinch valve; 24 VDC, 1/8" ID x 1/4" OD tubing
Includes
12" length of silicone tubing. Additional tubing sold separately. All valves have 15" 26-gauge PTFE coated lead wires
Key Features
  • Control flow in your tubing without direct contact between the fluid and the valve
  • Fast response time
  • Low-power consumption and high-cycle life
  • Excellent for sanitary and pharmaceutical applications
More About this Item
Choose from two-way NC, two-way NO, or three-way pinch valves. These are ideal where contamination is an issue, as the fluid only comes in contact with the tubing, not the valve. Tubing replacement is quick and easy, making these suitable for pharmaceutical or sanitary applications. Mounting clip or mounting flange available separately for increased versatility.